ENA447956225 Enamine

2-bromo-5-chloro-3-fluorobenzoic acid

  • MDL number MFCD24119836
  • Cas Number : 1529288-48-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203639

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203639
Availability details for
Quantity
  • Availability
  • Add to Cart