ENA447956226 Enamine

2-[3-(pyridin-2-ylsulfanyl)propyl]-2,3-dihydro-1H-isoindole-1,3-dione

  • MDL number MFCD02755477
  • Cas Number : 443120-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203660

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203660
Availability details for
Quantity
  • Availability
  • Add to Cart