ENA447956229 Enamine

(2R)-2-hydroxy-2,6,6-trimethylbicyclo[3.1.1]heptan-3-one

  • MDL number MFCD28397392
  • Cas Number : 1807938-57-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203716

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203716
Availability details for
Quantity
  • Availability
  • Add to Cart