ENA447956232 Enamine

1-ethyl-5-[2-(trimethylsilyl)ethynyl]-1H-pyrazole

  • MDL number MFCD26939972
  • Cas Number : 1707563-64-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203730

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203730
Availability details for
Quantity
  • Availability
  • Add to Cart