ENA447956234 Enamine

5H,6H,7H,8H-imidazo[1,5-a]pyridin-6-amine

  • MDL number MFCD18830060
  • Cas Number : 185796-62-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95
Storage temp. 0

Safety Information

msds https://www.enaminestore.com/catalog/EN300-145366
Availability details for
Quantity
  • Availability
  • Add to Cart