ENA447956235 Enamine

3-amino-N-[4-(dimethylamino)butyl]benzene-1-sulfonamide

  • MDL number MFCD15469582
  • Cas Number : 1292173-12-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203769

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203769
Availability details for
Quantity
  • Availability
  • Add to Cart