ENA447956240 Enamine

2-(4-chlorophenyl)cycloheptan-1-one

  • MDL number MFCD11870698
  • Cas Number : 107776-14-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203811

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203811
Availability details for
Quantity
  • Availability
  • Add to Cart