ENA447956243 Enamine

3-bromo-2-methyl-5-nitrobenzoic acid

  • MDL number MFCD20275367
  • Cas Number : 1269292-02-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203844

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203844
Availability details for
Quantity
  • Availability
  • Add to Cart