ENA447956247 Enamine

3-benzoylbenzene-1-sulfonyl chloride

  • MDL number MFCD10693749
  • Cas Number : 53558-61-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203863

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203863
Availability details for
Quantity
  • Availability
  • Add to Cart