ENA447956250 Enamine

1-(5-chlorothiophen-2-yl)-2-hydroxypropan-1-one

  • MDL number MFCD16165837
  • Cas Number : 1247434-48-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203867

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203867
Availability details for
Quantity
  • Availability
  • Add to Cart