ENA447956251 Enamine

1-[2-(4-chlorophenyl)-1,3-thiazol-5-yl]methanamine

  • MDL number MFCD14615241
  • Cas Number : 1206980-91-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203868
Availability details for
Quantity
  • Availability
  • Add to Cart