ENA447956252 Enamine

4-[2-(4-methylpiperazin-1-yl)ethoxy]phenol

  • MDL number MFCD11848903
  • Cas Number : 100221-01-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203887

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203887
Availability details for
Quantity
  • Availability
  • Add to Cart