ENA447956253 Enamine

2-[3-(2-methylpropoxy)phenyl]acetic acid

  • MDL number MFCD14695264
  • Cas Number : 916911-66-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203901

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203901
Availability details for
Quantity
  • Availability
  • Add to Cart