ENA447956254 Enamine

[3-(2-methylpropoxy)phenyl]methanol

  • MDL number MFCD11149531
  • Cas Number : 1021064-15-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203902

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203902
Availability details for
Quantity
  • Availability
  • Add to Cart