ENA447956255 Enamine

1-(chloromethyl)-3-(2-methylpropoxy)benzene

  • MDL number MFCD11115393
  • Cas Number : 1036486-25-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203903

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203903
Availability details for
Quantity
  • Availability
  • Add to Cart