ENA447956259 Enamine

N-methylisoquinoline-5-sulfonamide

  • MDL number MFCD11986510
  • Cas Number : 84468-20-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-203911

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-203911
Availability details for
Quantity
  • Availability
  • Add to Cart