ENA447956290 Enamine

5-(butan-2-yl)cyclohexane-1,3-dione

  • MDL number MFCD24088004
  • Cas Number : 57641-91-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204115

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204115
Availability details for
Quantity
  • Availability
  • Add to Cart