ENA447956291 Enamine

2-{[3-(trifluoromethyl)phenyl]methyl}pyrrolidine

  • MDL number MFCD12407016
  • Cas Number : 933687-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204123

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204123
Availability details for
Quantity
  • Availability
  • Add to Cart