ENA447956293 Enamine

4-methoxyoxane-4-carboxylic acid

  • MDL number MFCD12024537
  • Cas Number : 1010836-49-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204133

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204133
Availability details for
Quantity
  • Availability
  • Add to Cart