ENA447956294 Enamine

5-methyl-5-(1-methylcyclopropyl)imidazolidine-2,4-dione

  • MDL number MFCD20728986
  • Cas Number : 21231-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204141

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204141
Availability details for
Quantity
  • Availability
  • Add to Cart