ENA447956299 Enamine

4-(3,4-difluorophenyl)piperidine hydrochloride

  • MDL number MFCD04037205
  • Cas Number : 721958-67-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 255-256
Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204154

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204154
Availability details for
Quantity
  • Availability
  • Add to Cart