ENA447956303 Enamine

1-(3,4-diaminophenyl)ethan-1-one

  • MDL number MFCD12031488
  • Cas Number : 21304-39-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131-134
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204190

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204190
Availability details for
Quantity
  • Availability
  • Add to Cart