ENA447956306 Enamine

2-amino-2-(naphthalen-2-yl)acetic acid hydrochloride

  • MDL number MFCD06202323
  • Cas Number : 433292-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204199

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204199
Availability details for
Quantity
  • Availability
  • Add to Cart