ENA447956311 Enamine

7-methyl-5H,8H-imidazo[1,2-a]pyrimidin-5-one

  • MDL number MFCD09455049
  • Cas Number : 39567-76-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204231
Availability details for
Quantity
  • Availability
  • Add to Cart