ENA447956312 Enamine

1-(2-methyl-1,3-oxazol-4-yl)ethan-1-one

  • MDL number MFCD22071533
  • Cas Number : 81740-16-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204234

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204234
Availability details for
Quantity
  • Availability
  • Add to Cart