ENA447956314 Enamine

3-chloro-4-ethoxybenzonitrile

  • MDL number MFCD10666511
  • Cas Number : 916596-02-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204240

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204240
Availability details for
Quantity
  • Availability
  • Add to Cart