ENA447956320 Enamine

5-methyl-5-phenylazepan-2-one

  • MDL number MFCD24649796
  • Cas Number : 1803561-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204282

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204282
Availability details for
Quantity
  • Availability
  • Add to Cart