ENA447956323 Enamine

(5-bromopyridin-3-yl)(cyclopropyl)methanamine

  • MDL number MFCD18660969
  • Cas Number : 1270329-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204293

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204293
Availability details for
Quantity
  • Availability
  • Add to Cart