ENA447956324 Enamine

2-(piperidin-4-yl)ethan-1-amine dihydrochloride

  • MDL number MFCD03839805
  • Cas Number : 90000-31-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data 2HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204299

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204299
Availability details for
Quantity
  • Availability
  • Add to Cart