ENA447956326 Enamine

2-(oxolan-2-yl)pyrrolidine

  • MDL number MFCD02663538
  • Cas Number : 383127-91-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204317
Availability details for
Quantity
  • Availability
  • Add to Cart