ENA447956327 Enamine

3,5-difluoro-4-(trifluoromethyl)benzene-1-sulfonyl chloride

  • MDL number MFCD20369937
  • Cas Number : 1340212-34-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204325

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204325
Availability details for
Quantity
  • Availability
  • Add to Cart