ENA447956329 Enamine

2-bromocyclohexane-1-carbonitrile

  • MDL number MFCD24512494
  • Cas Number : 123804-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204334

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204334
Availability details for
Quantity
  • Availability
  • Add to Cart