ENA447956331 Enamine

5-[(tert-butoxy)carbonyl]pyrazine-2-carboxylic acid

  • MDL number MFCD26521436
  • Cas Number : 1192152-97-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204343

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204343
Availability details for
Quantity
  • Availability
  • Add to Cart