ENA447956333 Enamine

1,2,3-thiadiazole-4-carboxamide

  • MDL number MFCD01567032
  • Cas Number : 4100-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204367

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204367
Availability details for
Quantity
  • Availability
  • Add to Cart