ENA447956337 Enamine

1-(4-ethoxyphenyl)-2-methoxyethan-1-amine

  • MDL number MFCD09735326
  • Cas Number : 953715-26-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204392

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204392
Availability details for
Quantity
  • Availability
  • Add to Cart