ENA447956348 Enamine

methyl 2-cyano-2-methyl-2-phenylacetate

  • MDL number MFCD28397306
  • Cas Number : 79341-72-3
  • Molecular Weight : 189.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-156982

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-156982
Availability details for
Quantity
  • Availability
  • Add to Cart