ENA447956357 Enamine

tert-butyl N-(2-methyl-1,3-benzothiazol-6-yl)carbamate

  • MDL number MFCD21170409
  • Cas Number : 869096-35-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204467
Availability details for
Quantity
  • Availability
  • Add to Cart