ENA447956378 Enamine

(2-bromo-1-methoxyethyl)benzene

  • MDL number MFCD09952414
  • Cas Number : 13685-00-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204543
Availability details for
Quantity
  • Availability
  • Add to Cart