ENA447956379 Enamine

3-(1H-pyrazol-1-yl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

  • MDL number MFCD16995209
  • Cas Number : 2223054-07-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204544

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204544
Availability details for
Quantity
  • Availability
  • Add to Cart