ENA447956383 Enamine

2-bromo-4-methanesulfonylbenzene-1-sulfonyl chloride

  • MDL number MFCD28383818
  • Cas Number : 1603381-94-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204560

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204560
Availability details for
Quantity
  • Availability
  • Add to Cart