ENA447956387 Enamine

4-(3-methyl-1H-pyrazol-4-yl)piperidine

  • MDL number MFCD22068373
  • Cas Number : 1367915-64-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204580

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204580
Availability details for
Quantity
  • Availability
  • Add to Cart