ENA447956398 Enamine

2-amino-3-chloro-5-fluorobenzonitrile

  • MDL number MFCD18426017
  • Cas Number : 1263277-06-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204600

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204600
Availability details for
Quantity
  • Availability
  • Add to Cart