ENA447956403 Enamine

4-chloro-1-(2,2,2-trifluoroethyl)-1H-1,3-benzodiazol-2-amine

  • MDL number MFCD28383826
  • Cas Number : 1565106-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204605

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204605
Availability details for
Quantity
  • Availability
  • Add to Cart