ENA447956404 Enamine

3-(chloromethyl)-1lambda6-thiane-1,1-dione

  • MDL number MFCD21081233
  • Cas Number : 1481109-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204606

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204606
Availability details for
Quantity
  • Availability
  • Add to Cart