ENA447956411 Enamine

5-(methoxymethyl)-1,3-thiazol-2-amine

  • MDL number MFCD18816986
  • Cas Number : 920337-78-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204615

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204615
Availability details for
Quantity
  • Availability
  • Add to Cart