ENA447956412 Enamine

4-hydroxy-5-nitrobenzene-1,3-dicarbaldehyde

  • MDL number MFCD00839418
  • Cas Number : 70310-96-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204616

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204616
Availability details for
Quantity
  • Availability
  • Add to Cart