ENA447956415 Enamine

5-bromo-2-[(trifluoromethyl)sulfanyl]pyridine

  • MDL number MFCD19440970
  • Cas Number : 1204234-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204620

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204620
Availability details for
Quantity
  • Availability
  • Add to Cart