ENA447956419 Enamine

{1-[(benzylamino)methyl]cyclopropyl}methanol

  • MDL number MFCD23728920
  • Cas Number : 1546642-62-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204630

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204630
Availability details for
Quantity
  • Availability
  • Add to Cart