ENA447956435 Enamine

3-bromo-1-(1,2,3,4-tetrahydronaphthalen-1-yl)pyrrolidin-2-one

  • MDL number MFCD19187949
  • Cas Number : 1275893-77-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204688

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204688
Availability details for
Quantity
  • Availability
  • Add to Cart