ENA447956436 Enamine

3-(3-aminophenyl)-5-cyclopropyl-5-methylimidazolidine-2,4-dione

  • MDL number MFCD20097405
  • Cas Number : 1485279-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-204689

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-204689
Availability details for
Quantity
  • Availability
  • Add to Cart